Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Benzoin oxime, 98+%

Catalog No. aaa1063014
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 441-38-3
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00004501
InChI Key WAKHLWOJMHVUJC-FYWRMAATNA-N
Synonym 2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
PubChem CID 7057888
IUPAC Name (1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
SMILES O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 151°C to 154°C
Quantity 25 g
Beilstein 2051941
Sensitivity Light sensitive
Merck Index 14,1094
Formula Weight 227.26
Percent Purity ≥98%
Chemical Name or Material alpha-Benzoin oxime
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 207-127-2
RTECSNumber DI1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.