Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha/beta Tubulin Monoclonal Antibody (TU-10), Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. PIMA544130
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544130 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544130 Supplier Invitrogen™ Supplier No. MA544130
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes alpha, beta-tubulin heterodimer, a basic intracellular structural unit of microtubules. Alpha- and beta-tubulins form approximately 100 kDa tubulin heterodimer, a globular protein that polymerizes to form microtubules.

Tubulin protein is the major constituent of microtubules. Tubulin is a dimer of alpha and beta chains, which binds two molecules of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain. There are at least six isotypes of both alpha- and beta- tubulin in human cells, which are distinguished by slightly different amino acid sequences and encoded by a large, multigene family that has been highly conserved throughout evolution. Although the most important functions of microtubules in proliferative cells are through their actions as components of the mitotic spindle, they are also involved in many other essential functions throughout the cell cycle of both malignant and nonmalignant cells. Anti-microtubule agents including Vinca alkaloids and taxanes may disrupt many of these essential functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha/beta Tubulin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone TU-10
Concentration 0.1 mg/mL
Conjugate Alexa Fluor 488
Formulation TBS with 0.2% BSA and 15mM sodium azide
Gene TUBB
Gene Accession No. A2AQ07, P02550, P07437, P68369, P68370, Q71U36, Q767L7, Q9H4B7
Gene Alias 2810484G07Rik; Alpha-tubulin 1; Alpha-tubulin 3; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; beta 5-tubulin; beta Ib tubulin; CDCBM6; CSCSC1; Detyrosinated tubulin alpha-1A chain; FLJ30169; H2 ALPHA; hum-a-tub1; hum-a-tub2; LIS3; M(beta)1; M40; OK/SW-cl.56; Talpha1 alpha-tubulin; Tuba1; Tuba-1; TUBA1A; TUBA3; TUBB; TUBB1; TUBB2A; TUBB5; Tubulin alpha; tubulin alpha 1a; tubulin alpha 3; tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-3 chain; Tubulin B-alpha-1; tubulin beta; tubulin beta 1 class VI; tubulin beta chain; tubulin beta class I; tubulin beta-1 chain; Tubulin beta-5 chain; tubulin, alpha 1; tubulin, alpha 1A; tubulin, alpha, brain-specific; tubulin, beta 1 class VI; tubulin, beta 2A class IIa; tubulin, beta class I; tubulin, beta family 1; tubulin, beta polypeptide; α/β-tubulin
Gene Symbols TUBA1A, TUBB, TUBB1
Host Species Mouse
Immunogen Microtubule proteins from porcine brain (aa 1-444).
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100134983, 100135051, 203068, 22142, 545486, 64158, 679312, 733686, 7846, 81027
Target Species Human, Mouse, Rat, Monkey, Pig
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.