Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Bisabolol, 96%, Alfa Aesar™

Click to view available options
Quantity:
10g
50g
515-69-5
C15H26O
222.372
MFCD03846910
RGZSQWQPBWRIAQ-LSDHHAIUSA-N
bisabolol,alpha-bisabolol,+-alpha-bisabolol,d-alpha-bisabolol,.alpha.-bisabolol,+-1'r,2r-alpha-bisabolol,2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol,3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*,dsstox_cid_25964,dsstox_rid_81260
1549992
(2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
CC1=CCC(CC1)C(C)(CCC=C(C)C)O
515-69-5
C15H26O
222.372
MFCD03846910
RGZSQWQPBWRIAQ-LSDHHAIUSA-N
bisabolol,alpha-bisabolol,+-alpha-bisabolol,d-alpha-bisabolol,.alpha.-bisabolol,+-1'r,2r-alpha-bisabolol,2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol,3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*,dsstox_cid_25964,dsstox_rid_81260
1549992
(2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
CC1=CCC(CC1)C(C)(CCC=C(C)C)O
Applications
It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

Solubility
Soluble in alcohol, natural, mineral and synthetic oils. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Sensitivity Air Sensitive
Refractive Index 1.495
Density 0.93
Flash Point 135°C (275°F)
Quantity 10g
Beilstein 5733954
Merck Index 14,1241
Solubility Information Soluble in alcohol,natural,mineral and synthetic oils. Insoluble in water.
Formula Weight 222.37
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Bisabolol
Show More Show Less

EINECSNumber : 208-205-9

RTECSNumber : MJ9685000

TSCA : Yes

Recommended Storage : Ambient temperatures