Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ alpha Catenin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529247
Description
Recommended positive controls: A431, HeLa, HepG2, mouse brain, Rat2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
Specifications
alpha Catenin | |
Polyclonal | |
Unconjugated | |
CTNNA2 | |
102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI481747; AI988031; alpha E catenin; alpha E-catenin; Alpha N-catenin; alpha(E)-catenin; alpha(N)-catenin; alpha-catenin; alpha-catenin-related protein; alpha-E-catenin; alphaE-catenin; alpha-N-catenin; alphaN-catenin; cadherin associated protein; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; CAPR; CAP-R; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 1; catenin alpha 2; Catenin alpha1; catenin alpha-1; Catenin alpha2; Catenin alpha-2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; Catna; Catna1; Catna2; cdf; cerebellar deficient folia; chp; CT114; CTNNA1; Ctnna2; CTNR; FLJ36832; FLJ52416; HGNC:2509; MDPT2; Neural alpha-catenin; NY-REN-13 antigen; renal carcinoma antigen NY-REN-13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12385, 12386, 1495, 1496, 297357, 307505 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P26231, P26232, P35221, Q61301 | |
CTNNA1, CTNNA2 | |
Recombinant fragment corresponding to a region within amino acids 40 and 324 of Human alpha Catenin. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction