Learn More
Invitrogen™ alpha Catenin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120932
Description
Positive test controls include: BT-474, OVCAR3, 293T, MCF7, Mouse lung, Mouse heart, Mouse brain. The target is usually found in the following locations: Cell junction, Cell junction, Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, adherens junction, cytoskeleton. Immunogen sequence: ELDDSDFETE DFDVRSRTSV QTEDDQLIAG QSARAIMAQL PQEQKAKIAE QVASFQEEKS KLDAEVSKWD DSGNDIIVLA KQMCMIMMEM TDFTRGKGPL KNTSDVISAA KKIAEAGSRM DKLGRTIADH CPDSACKQDL LAYLQRIALY CHQLNICSKV KAEVQNLGGE LVVSGVDSAM SLIQAAKNLM NAVVQTVKAS YVASTKYQKS QGMASLNLPA VSWKMKAPEK KPLVKREKQD ETQTKIKRAS QKKHVNPVQA LSEFKAMDSI.
Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
Specifications
alpha Catenin | |
Polyclonal | |
Unconjugated | |
CTNNA2 | |
102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI481747; AI988031; alpha E catenin; alpha E-catenin; Alpha N-catenin; alpha(E)-catenin; alpha(N)-catenin; alpha-catenin; alpha-catenin-related protein; alpha-E-catenin; alphaE-catenin; alpha-N-catenin; alphaN-catenin; cadherin associated protein; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; CAPR; CAP-R; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 1; catenin alpha 2; Catenin alpha1; catenin alpha-1; Catenin alpha2; Catenin alpha-2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; Catna; Catna1; Catna2; cdf; cerebellar deficient folia; chp; CT114; CTNNA1; Ctnna2; CTNR; FLJ36832; FLJ52416; HGNC:2509; MDPT2; Neural alpha-catenin; NY-REN-13 antigen; renal carcinoma antigen NY-REN-13 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12385, 12386, 1495, 1496, 297357, 307505 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P26231, P26232, P35221, Q61301 | |
CTNNA1, CTNNA2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 637-906 of human alpha-Catenin (NP_0018942). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.