Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Chlorocinnamaldehyde, 90+%, Thermo Scientific™

Catalog No. AC393530250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18365-42-9
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.61
MDL Number MFCD00077440
InChI Key SARRRAKOHPKFBW-TWGQIWQCSA-N
Synonym alpha-chlorocinnamaldehyde, 2-chloro-3-phenylacrylaldehyde, 2-propenal, 2-chloro-3-phenyl, 2-chloro-3-phenyl-2-propenal, cinnamaldehyde, alpha-chloro, .alpha.-chlorocinnamaldehyde, 2z-2-chloro-3-phenylprop-2-enal, z-2-chloro-3-phenyl-2-propenal, a-chlorocinnamaldehyde, cinnamaldehyde, alpha-chloro-, z
PubChem CID 5899053
IUPAC Name (Z)-2-chloro-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=C(C=O)Cl

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 20°C to 22°C
Color Brown
Boiling Point 107°C to 108°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.6390 to 1.641
Linear Formula C6H5CH=C(Cl)CHO
Quantity 25 g
Formula Weight 166.61
Percent Purity 90+%
Physical Form Liquid After Melting
Chemical Name or Material α-chlorocinnamaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 242-237-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.