Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha COP I Antibody (SR1142), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32211650UL
Description
alpha COP I Monoclonal antibody specifically detects alpha COP I in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
alpha COP I | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1142 | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
alpha coat protein, Alpha-coat protein, Alpha-COP, coatomer protein complex, subunit alpha, coatomer subunit alpha, FLJ26320, HEPCOP, HEP-COPalpha-COP, xenin | |
A synthesized peptide derived from human alpha COP I (Uniprot #: P53621) | |
50 μg | |
Signal Transduction | |
1314 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction