Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ alpha COP Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142630
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_001091868.1; NP_004362.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms.
Specifications
alpha COP | |
Polyclonal | |
Unconjugated | |
COPA | |
AILJK; alpha coat protein; alpha-coat protein; Alpha-COP; AU040324; coatomer protein complex subunit alpha; coatomer subunit alpha; COPA; HEPCOP; HEP-COP; proxenin; xenin; Xenopsin-related peptide | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
1314 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P53621 | |
COPA | |
Peptide with sequence CTRASNLENST. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction