Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyano-4-hydroxycinnamic acid, Ultrapure MALDI Matrix, Thermo Scientific Chemicals

Catalog No. aaj67635exk
Click to view available options
Quantity:
5 x 10 mg

 

For use as a matrix in MALDI mass spectrometry analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28166-41-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00004204
InChI Key AFVLVVWMAFSXCK-YVMONPNESA-N
Synonym alpha-cyano-4-hydroxycinnamate, 2-cyano-4-hydroxycinnamate, 2-cyano-3-4-hydroxyphe-nyl acrylic acid, 2z-2-cyano-3-4-hydroxyphenyl prop-2-enoic acid
PubChem CID 9794103
IUPAC Name (Z)-2-cyano-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=C(C#N)C(=O)O)O

Specifications

Melting Point 250°C to 253°C
Color Yellow
Odor Odorless
Quantity 5 x 10 mg
Beilstein 3271427
Formula Weight 189.17
Physical Form Powder
Chemical Name or Material alpha-Cyano-4-hydroxycinnamic acid, Ultrapure MALDI Matrix
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 248-879-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.