Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%

Catalog No. AAA1206414
Click to view available options
Quantity:
5 g
25 g

CAS: 56401-20-8 | C6H13Na2O9P | 306.114 g/mol

alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56401-20-8
Molecular Formula C6H13Na2O9P
Molecular Weight (g/mol) 306.114
MDL Number MFCD00150839
InChI Key AQMGMJDBUKKFOG-PKXGBZFFSA-N
Synonym alpha-d-glucose-1-phosphate disodium salt tetrahydrate
PubChem CID 131855210
IUPAC Name sodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
SMILES C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]

Specifications

Melting Point ∼138°C
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 376.16 (304.10 Anhydrous)
Percent Purity 98%
Chemical Name or Material alpha-D-Glucose-1-phosphate disodium salt tetrahydrate
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 260-154-1
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.