Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ alpha-D-Glucose pentaacetate, 99%

Click to view available options
Quantity:
50g
250g
604-68-2
C16H22O11
390.341
MFCD00064071
LPTITAGPBXDDGR-LJIZCISZSA-N
alpha-d-glucose pentaacetate,a-d-glucopyranose pentaacetate,pentaacetyl-alpha-d-glucose,penta-o-acetyl-alpha-d-glucopyranose,unii-1cs424ns93,1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose,alpha-d-glucopyranose pentaacetate,2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate,glucopyranose, pentaacetate, .alpha.-d,a-d-glucose pentaacetate
2723636
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
604-68-2
C16H22O11
390.341
MFCD00064071
LPTITAGPBXDDGR-LJIZCISZSA-N
alpha-d-glucose pentaacetate,a-d-glucopyranose pentaacetate,pentaacetyl-alpha-d-glucose,penta-o-acetyl-alpha-d-glucopyranose,unii-1cs424ns93,1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose,alpha-d-glucopyranose pentaacetate,2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate,glucopyranose, pentaacetate, .alpha.-d,a-d-glucose pentaacetate
2723636
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
Melting Point 109°C to 113°C
Quantity 50g
Optical Rotation +102° (c=5 in Chloroform)
Beilstein 98852
Formula Weight 390.34
Percent Purity 99%
Chemical Name or Material alpha-D-Glucose pentaacetate