Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-D-(+)-Talose, 97%

Catalog No. AAB2122602
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2122602 0.5 g
AAB2122677 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2122602 Supplier Thermo Scientific Chemicals Supplier No. B2122602
Add to Cart

CAS: 2595-98-4 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2595-98-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00135834
InChI Key GZCGUPFRVQAUEE-UHFFFAOYNA-N
Synonym d-+-talose, aldehydo-d-talose, 2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal
PubChem CID 99459
ChEBI CHEBI:68461
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal
SMILES OCC(O)C(O)C(O)C(O)C=O

Specifications

Melting Point 133°C to 135°C
Quantity 0.5 g
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material alpha-D-(+)-Talose
EINECSNumber 219-996-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.