Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Desmuslin Antibody (OTI7H6), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271736AF532
Description
alpha Desmuslin Monoclonal specifically detects alpha Desmuslin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
alpha Desmuslin | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SYNM | |
Recombinant protein fragment corresponding to amino acids 329-580 of human alpha Desmuslin (NP_663780) produced in E.coli. | |
0.1 mL | |
23336.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI7H6 | |
Western Blot, Immunohistochemistry | |
desmuslin, DMNEC 1.4.99.1, EC 2.6.1.16, KIAA0353synemin beta, SYNDesmuslin, synemin, synemin alpha, synemin, intermediate filament protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction