Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    alpha Desmuslin Antibody (OTI7H6), FITC, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271736F
Description
alpha Desmuslin Monoclonal specifically detects alpha Desmuslin in Human samples. It is validated for Immunohistochemistry.Specifications
| alpha Desmuslin | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SYNM | |
| Recombinant protein fragment corresponding to amino acids 329-580 of human alpha Desmuslin (NP_663780) produced in E.coli. | |
| 0.1 mL | |
| 23336.0 | |
| Store at 4°C in the dark. | 
| Immunohistochemistry | |
| OTI7H6 | |
| Immunohistochemistry | |
| desmuslin, DMNEC 1.4.99.1, EC 2.6.1.16, KIAA0353synemin beta, SYNDesmuslin, synemin, synemin alpha, synemin, intermediate filament protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction