Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Dystroglycan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113820 Supplier Invitrogen™ Supplier No. PA5113820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPSPGSSAAP ATEVPDRDPE KSSEDDVYLH TVIPAVVVAA ILLIAGIIAM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Dystroglycan
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DAG1
Gene Accession No. Q62165
Gene Alias 156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9
Gene Symbols DAG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Mouse Dag1 (aa 721-770).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13138
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.