Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Dystroglycan Rat anti-Mouse,Human, Unconjugated, Clone: 3D7, MilliporeSigma™

Catalog No. MABN25152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN25152MI 25 μg
MABN25151MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN25152MI Supplier MilliporeSigma Supplier No. MABN251525UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Alpha Dystroglycan Monoclonal antibody specifically detects Alpha Dystroglycan in Mouse,Human samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha Dystroglycan
Applications Western Blot
Classification Monoclonal
Clone 3D7
Conjugate Unconjugated
Gene Accession No. NP_001263410
Gene Alias Alpha-DGP;Alpha-DG
Gene Symbols Dag1, Dag-1
Host Species Rat
Immunogen Murine alpha-dystroglycan-Fc-Fusion protein.
Molecular Weight of Antigen ∼97 kDa observed; 96.91 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 3D7 is a rat monoclonal antibody that detects human and murine alpha-dystroglycan. It targets an epitope within the C-terminal region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.