Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-ENaC Monoclonal Antibody (2G4), PerCP
GREENER_CHOICE

Catalog No. PIMA545351
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA545351 Supplier Invitrogen™ Supplier No. MA545351
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45351 was sufficient for detection of ENaC alpha in 15 μg of Mouse whole kidney lysate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 85kDa.

Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel's syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-ENaC
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2G4
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene SCNN1A
Gene Accession No. P37089, Q61180
Gene Alias alpha ENaC-2; alpha-ENaC; Alpha-NaCH; amiloride-sensitive epithelial sodium channel; amiloride-sensitive epithelial sodium channel alpha subunit; amiloride-sensitive sodium channel subunit alpha; amiloride-sensitive sodium channel subunit alpha 2; BESC2; ENaC; ENaC alpha; ENaCa; ENaCalpha; Epithelial Na(+) channel subunit alpha; epithelial sodium channel alpha subunit; FLJ21883; mENaC; nasal epithelial sodium channel alpha subunit; nonvoltage-gated sodium channel 1 subunit alpha; Renac; SCNEA; SCNN1; Scnn1a; sodium channel epithelial 1 alpha subunit; sodium channel, non voltage gated 1 alpha subunit; sodium channel, nonvoltage-gated 1 alpha; sodium channel, non-voltage-gated 1 alpha subunit; sodium channel, nonvoltage-gated, type I, alpha; sodium channel, nonvoltage-gated, type I, alpha polypeptide
Gene Symbols SCNN1A
Host Species Mouse
Immunogen Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20276, 25122
Target Species Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.