Learn More
Invitrogen™ alpha-ENaC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596353
Description
Immunogen sequence: LFGEYFSYPV SLNINLNSDK LVFPAVTICT LNPYRYPEIK EELEELDRIT EQTLFDLYKY SSFTTLVAGS RSRRDLRGTL PHPLQRLRVP PPPHGARRAR SVASSLRDNN PQVDWKDWKI G; Positive Samples: Mouse lung, Mouse intestine; Cellular Location: Apical cell membrane, Cell projection, Multi-pass membrane protein, cilium.
Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel's syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
Specifications
alpha-ENaC | |
Polyclonal | |
Unconjugated | |
SCNN1A | |
alpha ENaC-2; alpha-ENaC; Alpha-NaCH; amiloride-sensitive epithelial sodium channel; amiloride-sensitive epithelial sodium channel alpha subunit; amiloride-sensitive sodium channel subunit alpha; amiloride-sensitive sodium channel subunit alpha 2; BESC2; ENaC; ENaC alpha; ENaCa; ENaCalpha; Epithelial Na(+) channel subunit alpha; epithelial sodium channel alpha subunit; FLJ21883; mENaC; nasal epithelial sodium channel alpha subunit; nonvoltage-gated sodium channel 1 subunit alpha; Renac; SCNEA; SCNN1; Scnn1a; sodium channel epithelial 1 alpha subunit; sodium channel, non voltage gated 1 alpha subunit; sodium channel, nonvoltage-gated 1 alpha; sodium channel, non-voltage-gated 1 alpha subunit; sodium channel, nonvoltage-gated, type I, alpha; sodium channel, nonvoltage-gated, type I, alpha polypeptide | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20276, 25122, 6337 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
3.12 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P37088, P37089, Q61180 | |
SCNN1A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 105-225 of human SCNN1A (NP_0010291). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.