Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Endosulfine Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300273UV
Description
alpha Endosulfine Polyclonal specifically detects alpha Endosulfine in Human samples. It is validated for Western Blot, ELISA.Specifications
alpha Endosulfine | |
Polyclonal | |
Western Blot, ELISA | |
alpha-endosulfine, ARPP-19eMGC4319, endosulfine alpha, MGC78563, MGC8394 | |
Produced in rabbits immunized with purified, recombinant alpha Endosulfine (O43768-1; Met1-Glu121). | |
0.1 mL | |
Neuroscience | |
2029 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction