Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Fetoprotein/AFP Antibody (5H7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010072G
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
alpha-Fetoprotein/AFP | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AFP | |
Full length protein (purified) (Human). | |
0.2 mL | |
Cancer, Cancer Stem Cells, GPCR, Stem Cell Markers, Tumor Biomarkers | |
174 | |
Human | |
Purified |
ELISA | |
5H7 | |
ELISA, Sandwich ELISA | |
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
No cross-reaction with human albumin. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction