Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Fetoprotein/AFP Antibody (AFP-11), Alexa Fluor™ 405

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500524AF405
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
alpha-Fetoprotein/AFP | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AFP | |
purified alpha-Fetoprotein | |
0.1 mL | |
Cancer, Cancer Stem Cells, GPCR, Stem Cell Markers, Tumor Biomarkers | |
174 | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
AFP-11 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This antibody (clone AFP-11) reacts with human alpha-Fetoprotein (AFP), a 70 kDa oncofetal antigen. AFP is a major fetal plasma protein, but is not present in healthy adult tissues. Elevated AFP concentrations in adult plasma may be an early marker of hepatocellular carcinoma or teratoblastoma, while high concentrations in amniotic fluid may indicate severe congenital defects of a fetus. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction