Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Fetoprotein/AFP Antibody (C3), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234577AF750
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human, Porcine, Canine, Monkey, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA, Flow (Intracellular), Knockout Validated.Specifications
alpha-Fetoprotein/AFP | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
AFP | |
Alpha feto protein (AFP) Purified from serum of a hepatoma patient (Uniprot: P02771) | |
0.1 mL | |
Primary | |
It recognizes an oncofetal glycoprotein with a single chain of 70kDa, which is identified as alpha-fetoprotein (AFP) (ISOBM TD-2 workshop). This monoclonal antibody is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
C3 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated | |
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
Mouse | |
Protein A or G purified | |
Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers | |
174.0 | |
Human, Pig, Canine, Primate, Bovine, Mouse, Rat | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction