Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Fetoprotein/AFP Rabbit anti-Human, Clone: AF5, Novus Biologicals™

Rabbit Monoclonal Antibody
$234.19 - $615.90
Specifications
| Antigen | alpha-Fetoprotein/AFP |
|---|---|
| Clone | AF5 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| alpha-Fetoprotein/AFP | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers | |
| PBS | |
| Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
| IgG κ | |
| Protein A purified |
| AF5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P02771 | |
| 174 | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title