Learn More
Alpha Fodrin Antibody (3D7), Alexa Fluor™ 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP192689AF405
Catalog No. NBP192689W
Only null left
Add to Cart
Description
Description
Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Alpha Fodrin
Monoclonal
Alexa Fluor 405
Q13813
SPTAN1
Recombinant construct (expressed in and purified from E. coli) containing the 19th and 20th spectrin repeats and C-terminal EF hand domains of human alpha Fodrin (amino acids 2086-2447) [UniProt# Q13813].
Protein G purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
3D7
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2
Mouse
240 kDa
0.1 mL
Neuroscience
6709
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only