Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Antibody (3D7), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192689DL594
Description
Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Alpha Fodrin | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 3D7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
| Recombinant construct (expressed in and purified from E. coli) containing the 19th and 20th spectrin repeats and C-terminal EF hand domains of human alpha Fodrin (amino acids 2086-2447) [UniProt# Q13813]. | |
| 0.1 mL | |
| Neuroscience | |
| 6709 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction