Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Antibody (3D7), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Alpha Fodrin |
|---|---|
| Clone | 3D7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:3000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP192689SS
![]() |
Novus Biologicals
NBP192689SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP192689
![]() |
Novus Biologicals
NBP192689 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Alpha Fodrin | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
| SPTAN1 | |
| IgG1 | |
| Immunogen affinity purified | |
| 240 kDa |
| 3D7 | |
| Western Blot 1:3000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q13813 | |
| 6709 | |
| Recombinant construct (expressed in and purified from E. coli) containing the 19th and 20th spectrin repeats and C-terminal EF hand domains of human alpha Fodrin (amino acids 2086-2447) [UniProt# Q13813]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title