Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060441AF350
Description
Alpha Fodrin Polyclonal antibody specifically detects Alpha Fodrin in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Alpha Fodrin | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
The immunogen recognized by this antibody maps to a region between residue 1000 and 1050 of human spectrin, alpha, non-erythrocytic 1 using the numbering given in entry NP_003118.1 | |
0.1 mL | |
Neuroscience | |
6709 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction