Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Antibody (SPTAN1/3505), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314184AF750
This item is not returnable.
View return policy
Description
Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Alpha Fodrin | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPTAN1/3505 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
Recombinant fragment of human Alpha Fodrin protein (around aa2351-2475) (exact sequence is proprietary)) (Uniprot: Q13813) | |
0.1 mL | |
Neuroscience | |
6709 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction