Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Mouse anti-Human, Clone: SPTAN1/3352, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30726720UG
Description
Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Alpha Fodrin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
Recombinant fragment of human Alpha Fodrin protein (around aa 2351-2475) (exact sequence is proprietary)) | |
20 μg | |
Neuroscience | |
6709 | |
Store at 4°C. | |
IgG2c κ |
Immunohistochemistry (Paraffin), Peptide Array | |
SPTAN1/3352 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction