Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Fodrin Mouse anti-Human, Clone: SPTAN1/3505, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
| Antigen | Alpha Fodrin |
|---|---|
| Clone | SPTAN1/3505 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Alpha Fodrin | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
| Recombinant fragment of human Alpha Fodrin protein (around aa2351-2475) (exact sequence is proprietary)) (Uniprot: Q13813) | |
| Primary | |
| Store at 4°C. |
| SPTAN1/3505 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q13813 | |
| 6709 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title