Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ alpha Galactosidase Monoclonal Antibody (C8)
Mouse Monoclonal Antibody
$213.50 - $561.50
Specifications
Antigen | alpha Galactosidase |
---|---|
Clone | C8 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541589
|
Invitrogen™
MA541589 |
200 μL |
Each for $561.50
|
|
|||||
PIMA543975
|
Invitrogen™
MA543975 |
20 μL |
Each for $213.50
|
|
|||||
Description
alpha Galactosidase Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predominantly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.Specifications
alpha Galactosidase | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P06280 | |
2717 | |
Recombinant protein Galactosidase Alpha. The antigen corresponds to amino acid range 81-429 of the target protein. | |
Antibody |
C8 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
GLA | |
Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; Melibiase | |
GLA | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title