Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Internexin Antibody (2E3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300140B
Description
alpha-Internexin Monoclonal antibody specifically detects alpha-Internexin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
alpha-Internexin | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2000 | |
Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1 | |
Purified recombinant rat alpha Internexin expressed in and purified from E. coli [UniProt# P23565] | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
9118 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction