Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Internexin Antibody (529724) [mFluor Violet 610 SE], Novus Biologicals™
SDP

Catalog No. NB655700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB655700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB655700 Supplier Novus Biologicals Supplier No. FAB4844MFV610
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Internexin Monoclonal antibody specifically detects alpha-Internexin in Human samples. It is validated for Immunohistochemistry,Western Blot

Specifications

Antigen alpha-Internexin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 529724
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1
Host Species Mouse
Immunogen E. coli-derived recombinant human alpha -Internexin, Val230-Glu450, Accession # Q16352
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9118
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.