Learn More
Invitrogen™ alpha Internexin Monoclonal Antibody (1D2)
Description
In Western blot, a band at ~66 kDa is detected. The epitope recognized by MA1-4709 (C-terminal non-helical extension of the protein) is unusually resistant to aldehyde fixation, so formalin fixation of histological sections is recommended.
Specifications
Specifications
| Antigen | alpha Internexin |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1D2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10mM sodium azide |
| Gene | INA |
| Gene Accession No. | P23565, P46660, Q16352 |
| Gene Alias | 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.