Learn More
Invitrogen™ alpha Internexin Monoclonal Antibody (1D2)

Description
alpha Internexin Monoclonal Antibody for Western Blot, IHC (Free), ICC/IF
Specifications
Specifications
| Antigen | alpha Internexin |
| Applications | Immunohistochemistry (Free Floating), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1D2 |
| Concentration | 1 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.035% sodium azide |
| Gene | INA |
| Gene Accession No. | P23565, P46660, Q16352 |
| Gene Alias | 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.