Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Internexin Monoclonal Antibody (2E3)
GREENER_CHOICE

Catalog No. AMSA6726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMSA6726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMSA6726 Supplier Invitrogen™ Supplier No. MA316726
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The epitope recognized by the 2E3 clone is in the C-terminal non-helical extension of the protein and is unusually resistant to aldehyde fixation, so this antibody is ideal for studies of formalin-fixed paraffin-embedded histological sections.

alpha-Internexin is a Class IV intermediate filament subunit expressed very early in neuronal development. It is related to but distinct from the better known neurofilament triplet proteins, NF-L, NF-M and NF-H, having similar protein sequence motifs and a similar intron organization. Many classes of mature neurons contain a-internexin in addition to NF-L, NF-M and NF-H. In some mature neurons a-internexin is the only neurofilament subunit expressed. Antibodies to internexin are therefore unique probes to study and classify neuronal types and follow their processes in sections and in tissue culture. In addition the very early developmental expression of a-internexin means its presence is an early and convenient diagnostic feature of neuronal progenitors cells and other cell committed to the neuronal lineage. The use of antibodies to this protein in the study of brain tumors has not been examined to date, but is likely to be of interest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Internexin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.035% sodium azide
Gene INA
Gene Accession No. P23565, P46660, Q08DH7, Q16352
Gene Alias 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1
Gene Symbols INA
Host Species Mouse
Immunogen Purified recombinant rat alpha Internexin expressed in and purified from E. coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100153400, 226180, 24503, 532236, 9118
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.