Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ alpha Internexin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585789
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (83%). Positive Control: mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
alpha-Internexin is a Class IV intermediate filament subunit expressed very early in neuronal development. It is related to but distinct from the better known neurofilament triplet proteins, NF-L, NF-M and NF-H, having similar protein sequence motifs and a similar intron organization. Many classes of mature neurons contain a-internexin in addition to NF-L, NF-M and NF-H. In some mature neurons a-internexin is the only neurofilament subunit expressed. Antibodies to internexin are therefore unique probes to study and classify neuronal types and follow their processes in sections and in tissue culture. In addition the very early developmental expression of a-internexin means its presence is an early and convenient diagnostic feature of neuronal progenitors cells and other cell committed to the neuronal lineage. The use of antibodies to this protein in the study of brain tumors has not been examined to date, but is likely to be of interest.
Specifications
alpha Internexin | |
Polyclonal | |
Unconjugated | |
INA | |
66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
226180, 24503 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.24 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P23565, P46660 | |
INA | |
Recombinant protein encompassing a sequence within the C-terminus region of mouse alpha Internexin. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction