Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-L-arabinofuranosidase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984464
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4464 Supplier Bioss Supplier No. BS2045R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASD1 (Atrial Septal Defect 1) is a Genetic Locus. Diseases associated with ASD1 include Atrial Septal Defect 1 and Atrial Heart Septal Defect.

Specifications

Antigen alpha-L-arabinofuranosidase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ASD1
Gene Symbols ASD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Prunus persica alpha-L-arabinofuranosidase .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 431
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.