Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Lipoic Acid Dietary Supplement, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18600434
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

L1506, 1077-28-7, C8H14O2S2

Spectrum™ Chemical Alpha Lipoic Acid, USP Dietary Supplement is used for diabetes and nerve-related symptoms of diabetes including burning, pain, and numbness in the legs and arms. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
IUPAC Name 5-(1,2-dithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.2%
Melting Range 60-62°C
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation -1.0° to +1.0°
Percent Purity 99 to 101%
Grade USP Dietary Supplement
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.