Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha-mannosidase II Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP189748
Description
Alpha-mannosidase II Polyclonal specifically detects Alpha-mannosidase II in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Alpha-mannosidase II | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
alpha-mannosidase 2, AMan II, EC 3.2.1, EC 3.2.1.114, Golgi alpha-mannosidase II, golgi integral membrane protein 7, GOLIM7, Man II, MANA2MANII, Mannosidase alpha class 2A member 1, mannosidase, alpha type II, mannosidase, alpha, class 2A, member 1, Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase | |
Rabbit | |
Affinity Purified | |
RUO | |
4124 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MAN2A1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LVVYNPLEQDRISLVSVYVSSPTVQVFSASGKPVEVQVSAVWDTANTISETAYEISFRAHIPPLGLKVYKILESASSNSHLADYVLYKNKVEDSGIFTIKNMINTEEGITLENSFVLLRFDQTGLMKQMMTKED | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction