Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Alpha-mannosidase II Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC22186 and based on accession number NM_2372. The protein contains a C-terminal DDK tag. |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens mannosidase, alpha, class 2A, member 1 (MAN2A1), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
