Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Methylacyl-CoA Racemase/AMACR Antibody (AMACR/1723) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | alpha-Methylacyl-CoA Racemase/AMACR |
---|---|
Clone | AMACR/1723 |
Concentration | 1 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254501100
![]() |
Novus Biologicals
NBP254501100UG |
100 μg |
Each for $646.00
|
|
|||||
NB254501200
![]() |
Novus Biologicals
NBP254501200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
alpha-Methylacyl-CoA Racemase/AMACR Monoclonal specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
alpha-Methylacyl-CoA Racemase/AMACR | |
1 mg/mL | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
PBS with No Preservative | |
23600 | |
Recombinant human full-length AMACR protein | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
AMACR/1723 | |
Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM | |
AMACR | |
IgG | |
Protein A or G purified | |
54 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title