Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Methylacyl-CoA Racemase/AMACR Antibody (AMACR/1723), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
| Antigen | alpha-Methylacyl-CoA Racemase/AMACR |
|---|---|
| Clone | AMACR/1723 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25339020
![]() |
Novus Biologicals
NBP25339020UG |
20 μg |
Each for $268.00
|
|
|||||
NB253390100
![]() |
Novus Biologicals
NBP253390100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253390200
![]() |
Novus Biologicals
NBP253390200UG |
200 μg | N/A | N/A | N/A | ||||
Description
alpha-Methylacyl-CoA Racemase/AMACR Monoclonal specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| alpha-Methylacyl-CoA Racemase/AMACR | |
| 0.2 mg/mL | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 23600 | |
| Recombinant human full-length AMACR protein | |
| Primary | |
| Store at 4C. |
| AMACR/1723 | |
| Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM | |
| AMACR | |
| IgG | |
| Protein A or G purified | |
| 54 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title