Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Methylcinnamic acid, 99%

Catalog No. AAL0080614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0080614 25 g
AAL0080606 5 g
AAL0080622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0080614 Supplier Thermo Scientific Chemicals Supplier No. L0080614
Add to Cart

CAS: 1199-77-5 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1199-77-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00002652
InChI Key XNCRUNXWPDJHGV-BQYQJAHWSA-N
Synonym alpha-methylcinnamic acid, 2-methyl-3-phenylacrylic acid, 2e-2-methyl-3-phenylprop-2-enoic acid, a-methylcinnamic acid, 2e-2-methyl-3-phenylacrylic acid, .alpha.-methylcinnamic acid, e-2-methyl-3-phenylacrylic acid, 2-propenoic acid, 2-methyl-3-phenyl, alpha-methyl cinnamic acid, e-2-methyl-3-phenylprop-2-enoic acid
PubChem CID 637817
IUPAC Name (E)-2-methyl-3-phenylprop-2-enoic acid
SMILES C\C(=C/C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 78°C to 82°C
Boiling Point 288°C
Quantity 25 g
Beilstein 2042544
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material alpha-Methylcinnamic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-847-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.