Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Methylstyrene, 99%, stabilized, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
Packaging:
Glass bottle
Plastic drum
98-83-9,98-29-3
C9H10
118.18
MFCD00008859
XYLMUPLGERFSHI-UHFFFAOYSA-N
alpha-methylstyrene,2-phenylpropene,2-phenyl-1-propene,isopropenylbenzene,benzene, 1-methylethenyl,2-phenylpropylene,1-methyl-1-phenylethylene,alpha-methylstyrol,as-methylphenylethylene,1-phenyl-1-methylethylene
7407
CHEBI:35060
prop-1-en-2-ylbenzene
CC(=C)C1=CC=CC=C1
98-83-9,98-29-3
C9H10
118.18
MFCD00008859
XYLMUPLGERFSHI-UHFFFAOYSA-N
alpha-methylstyrene,2-phenylpropene,2-phenyl-1-propene,isopropenylbenzene,benzene, 1-methylethenyl,2-phenylpropylene,1-methyl-1-phenylethylene,alpha-methylstyrol,as-methylphenylethylene,1-phenyl-1-methylethylene
7407
CHEBI:35060
prop-1-en-2-ylbenzene
CC(=C)C1=CC=CC=C1
Viscosity 0.94 mPa.s (20°C)
Specific Gravity 0.909
Stabilizer 15ppm tert.-butylcatechol
Melting Point -23.0°C
Density 0.9090g/mL
Color Colorless
Flash Point 45°C
Refractive Index 1.5370 to 1.5390
Infrared Spectrum Authentic
Quantity 100g
Boiling Point 165.0°C to 169.0°C
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5C(CH3)=CH2
Beilstein 05, 484
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and benzene, soluble in aromatic hydrocarbons and other organic, solvents
Formula Weight 118.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material α-Methylstyrene, Stabilized
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Suspected of

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contami

GHS Signal Word: Danger

EINECSNumber : 202-705-