Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Methylstyrene, 99%, stabilized

Catalog No. p-3779195
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 98-83-9 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-83-9, 98-29-3
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008859
InChI Key XYLMUPLGERFSHI-UHFFFAOYSA-N
Synonym alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene
PubChem CID 7407
ChEBI CHEBI:35060
IUPAC Name prop-1-en-2-ylbenzene
SMILES CC(=C)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -23.0°C
Color Colorless
Density 0.9090g/mL
Boiling Point 165.0°C to 169.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(CH3)=CH2
Refractive Index 1.5370 to 1.5390
Quantity 100 g
Beilstein 05, 484
Specific Gravity 0.909
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and benzene, soluble in aromatic hydrocarbons and other organic, solvents
Stabilizer 15ppm tert.-butylcatechol
Viscosity 0.94 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material α-Methylstyrene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Suspected of
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contami
EINECSNumber 202-705-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.