Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha N-Catenin Monoclonal Antibody (T.44.7)
GREENER_CHOICE

Catalog No. PIMA514963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514963 Supplier Invitrogen™ Supplier No. MA514963
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other -alpha-catenin family members.

The catenins (alpha, beta, and gamma) are cytoplasmic proteins found in varying abundance in many developing and adult tissues. Catenins bind directly or indirectly to the conserved cytoplasmic tail domain of the cell adhesion cadherins. The association of catenins to cadherins produces a complex, which is linked to the actin filament network. Catenins/cadherin complexes play an important role in mediating cell adhesion, transduction of cell-cell contact positional signals to the cell interior, and may play a crucial role in cell differentiation. Alpha-Catenin (CAP102, 102 kDa), originally described as an E-cadherin associated protein, has been shown to associate with other members of the cadherin family members, N-cadherin and P-cadherin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha N-Catenin
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone T.44.7
Concentration 135 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
Gene CTNNA2
Gene Accession No. P26232, Q61301
Gene Alias AI481747; alpha N-catenin; alpha(N)-catenin; alpha-catenin-related protein; alpha-N-catenin; cadherin associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAPR; CAP-R; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 2; catenin alpha 2; catenin alpha-2; Catna; Catna2; cdf; cerebellar deficient folia; chp; CT114; CTNNA2; CTNR; DKFZp686H02198; OTTHUMP00000216460; OTTHUMP00000216462
Gene Symbols CTNNA2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the carboxy-terminal sequence of human a-N-Catenin.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12386, 1496, 297357
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.