Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-N-terminal Methyltransferase 1A/METTL11A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7159SP
This item is not returnable.
View return policy
Description
alpha-N-terminal Methyltransferase 1A/METTL11A Polyclonal specifically detects alpha-N-terminal Methyltransferase 1A/METTL11A in Human samples. It is validated for Western Blot.Specifications
alpha-N-terminal Methyltransferase 1A/METTL11A | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AD-003, alpha N-terminal protein methyltransferase 1A, C9orf32, chromosome 9 open reading frame 32, EC 2.1.1.-, methyltransferase like 11A, Methyltransferase-like protein 11A, NRMT, N-terminal RCC1 methyltransferase, NTM1A, NTMT1, X-Pro-Lys N-terminal protein methyltransferase 1A | |
Sheep | |
Affinity Purified | |
RUO | |
28989 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
Q9BV86 | |
NTMT1 | |
E. coli-derived recombinant human alpha-N-terminal Methyltransferase 1A/METTL11A Thr2-Arg223 Accession # Q9BV86 | |
25 μg | |
Primary | |
Detects human alpha-N-terminal Methyltransferase 1A/METTL11A in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction