Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Naphtholphthalein

Catalog No. p-7051448
Click to view available options
Quantity:
1 g
5 g
25 g

Indicator in 0.1% or 0.04% alcohol solution | CAS: 596-01-0 | C28H18O4 | 418.448 g/mol

Indicator in 0.1% or 0.04% alcohol solution.α-Naphtholphthalein is used as a pH indicator and operates in the pH ranges between 7.3 and 8.7. It is also used in the designing of optical carbon dioxide sensor based on fluorescence intensity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-01-0
Molecular Formula C28H18O4
Molecular Weight (g/mol) 418.448
MDL Number MFCD00036202
InChI Key HQHBAGKIEAOSNM-UHFFFAOYSA-N
Synonym alpha-naphtholphthalein, naphtholphthalein, 1-naphtholphthalein, a-naphtholphthalein, 3,3-bis 4-hydroxynaphthalen-1-yl isobenzofuran-1 3h-one, unii-2qeg1706nn, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-1-naphthalenyl, naphtholphthalein blue, alpha-naphtholphtalein, 3,3-bis 4-hydroxy-1-naphthyl phthalide
PubChem CID 68993
IUPAC Name 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O

Specifications

Melting Point 238°C to 240°C
Color Brown
Quantity 1 g
Merck Index 14,6389
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 418.45
Physical Form Powder
Chemical Name or Material alpha-Naphtholphthalein
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 209-875-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.