Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Pinene Oxide 95.0+%, TCI America™
SDP

Catalog No. P1362250ML
Click to view available options
Quantity:
25 mL
250 mL

Chemical Identifiers

CAS 1686-14-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00066955
InChI Key NQFUSWIGRKFAHK-UHFFFAOYSA-N
Synonym 2,3-Epoxypinane
PubChem CID 91508
ChEBI CHEBI:29060
SMILES CC1(C2CC1C3(C(C2)O3)C)C

Specifications

Color Yellow
Quantity 250 mL
UN Number 2810
Formula Weight 152.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Pinene Oxide
EINECSNumber (4)-0616
RTECSNumber TK4565000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.